1-Chloro-3,5-di(4-chlorbenzoyl)-2-deoxy-D-ribose 90%

Supplier: Ambeed
Warning

A1003248-100G A1003248-1G A1003248-25G A1003248-5G
76646-060EA 179.3 USD
76646-060 76646-062 76646-064 76646-066
1-Chloro-3,5-di(4-chlorbenzoyl)-2-deoxy-D-ribose 90%
1-Chloro-3,5-di(4-chlorbenzoyl)-2-deoxy-D-ribose

Precaución: for Research Use Only

Formula: C₁₉H₁₅Cl₃O₅
MW: 429.6784 g/mol
MDL Number: MFCD00672145
CAS Number: 3601-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR