5-Chloro-3-methylanthranilic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-chloro-3-methylbenzoic acid
A1003242-100G
A1003242-500G
76646-056EA
36.08
USD
76646-056
76646-058
5-Chloro-3-methylanthranilic acid 97%
5-Chloro-3-methylanthranilic acid
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₂ MW: 185.6076 g/mol |
MDL Number:
MFCD02358895 CAS Number: 20776-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...