11-Bromoundecyl 4-acetoxy-3-methoxybenzoate 97%

Supplier: Ambeed
Warning

A1003228-1G A1003228-250MG A1003228-5G
76646-042EA 10.61 USD
76646-042 76646-044 76646-046 76646-048
11-Bromoundecyl 4-acetoxy-3-methoxybenzoate 97%
11-Bromoundecyl 4-acetoxy-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₂₁H₃₁BrO₅
CAS Number: 2102353-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR