(Triphenylmethyl)thionyl Imide 98%

Supplier: Ambeed
Danger

Synonyms: Tritylthionyl Imide, TrNSO

A1003202-5G A1003202-1G
76646-038EA 221.73 USD
76646-038 77570-020
(Triphenylmethyl)thionyl Imide 98%
(Triphenylmethyl)thionyl Imide

Precaución: for Research Use Only

Formula: C₁₉H₁₅NOS
MW: 305.40 g/mol
MDL Number: MFCD32062442
CAS Number: 503596-47-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR