6-Chloro-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A1003200-100MG A1003200-1G A1003200-250MG A1003200-5G
76646-030EA 20.16 USD
76646-030 76646-032 76646-034 76646-036
6-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
6-Chloro-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀ClN
MW: 167.6354 g/mol
MDL Number: MFCD06739156
CAS Number: 33537-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR