3-Bromo-5-fluoro-4-iodoaniline 95%
Supplier: Ambeed
|
Warning
|
A1003188-100MG
A1003188-50MG
76646-026EA
184.6
USD
76646-026
76646-028
3-Bromo-5-fluoro-4-iodoaniline 95%
3-Bromo-5-fluoro-4-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrFIN MW: 315.9095 g/mol |
CAS Number:
2090537-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...