4-Phenoxyaniline 97%, (Chunks or pellets or flakes)

Supplier: Ambeed
Warning

Synonyms: 1-Amino-4-phenoxybenzol

A1003174-100G A1003174-25G A1003174-500G
76646-020EA 35.01 USD
76646-020 76646-022 76646-024
4-Phenoxyaniline 97%, (Chunks or pellets or flakes)
4-Phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00007862
CAS Number: 139-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR