5-Methylthiazole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A1003164-5G A1003164-250MG
76646-016EA 235.52 USD
76646-016 77619-494
5-Methylthiazole-2-carbaldehyde 95%
5-Methylthiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C5H5NOS
MW: 127.1643 g/mol
MDL Number: MFCD08060733
CAS Number: 13838-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR