tert-Butyl 4-(2-Amino-1-hydroxyethyl)piperidine-1-carboxylate 4-methylbenzenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A1003158-250MG
A1003158-5G
76646-012EA
38.2
USD
76646-012
76646-014
tert-Butyl 4-(2-Amino-1-hydroxyethyl)piperidine-1-carboxylate 4-methylbenzenesulfonate 97%
tert-Butyl 4-(2-Amino-1-hydroxyethyl)piperidine-1-carboxylate 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₉H₃₂N₂O₆S MW: 416.5322 g/mol |
MDL Number:
MFCD32854353 CAS Number: 2682112-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...