2-Bromo-4-methyl-1-nitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-nitrotoluene

A1003151-1G A1003151-5G
76646-008EA 28.65 USD
76646-008 76646-010
2-Bromo-4-methyl-1-nitrobenzene 95%
2-Bromo-4-methyl-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD07363742
CAS Number: 40385-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR