4,4'-Diethynyl-2,2'-bipyridine 98%

Supplier: Ambeed
Warning

A1323847-100MG A1323847-1G A1323847-5G A1323847-250MG
76657-860EA 16 USD
76657-860 76657-862 76657-864 77867-260
4,4'-Diethynyl-2,2'-bipyridine 98%
4,4'-Diethynyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₄H₈N₂
MW: 204.2267 g/mol
MDL Number: MFCD22572873
CAS Number: 133810-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR