Oxetan-3-ol 93%

Supplier: Ambeed
Danger

A1003128-100G A1003128-1G A1003128-25G A1003128-5G A1003128-10G
76645-996EA 106 USD
76645-996 76645-998 76646-000 76646-002 77570-018
Oxetan-3-ol 93%
Oxetan-3-ol

Precaución: for Research Use Only

Formula: C₃H₆O₂
MDL Number: MFCD09056790
CAS Number: 7748-36-9
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR