(E)-Phenethyl 3-phenylacrylate 99%

Supplier: Ambeed
Warning

A1003015-100G A1003015-25G A1003015-5G
76645-990EA 20.16 USD
76645-990 76645-992 76645-994
(E)-Phenethyl 3-phenylacrylate 99%
(E)-Phenethyl 3-phenylacrylate

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₂
MW: 252.3077 g/mol
MDL Number: MFCD00022050
CAS Number: 63238-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR