tert-Butyl 3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A1002807-100MG A1002807-1G A1002807-250MG
76645-976EA 24.41 USD
76645-976 77619-490 77619-492
tert-Butyl 3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate hydrochloride 97%
tert-Butyl 3-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₃ClN₂O₄
MW: 294.7750 g/mol
MDL Number: MFCD29065946
CAS Number: 1858241-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR