(S)-2-(4-(Methylsulfonyl)piperazin-1-yl)propan-1-ol 98% +(97%ee)

Supplier: Ambeed
Warning

A1002591-100G A1002591-1G A1002591-25G A1002591-500G A1002591-5G
76645-964EA 694.89 USD
76645-964 76645-966 76645-968 76645-970 76645-972
(S)-2-(4-(Methylsulfonyl)piperazin-1-yl)propan-1-ol 98% +(97%ee)
(S)-2-(4-(Methylsulfonyl)piperazin-1-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₃S
MW: 222.3051 g/mol
CAS Number: 2381259-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR