9-(Naphthalen-1-yl)-10-(4-(naphthalen-2-yl)phenyl)anthracene 97%

Supplier: Ambeed
Warning

A1002587-1G A1002587-250MG A1002587-5G
76645-958EA 43.5 USD
76645-958 76645-960 76645-962
9-(Naphthalen-1-yl)-10-(4-(naphthalen-2-yl)phenyl)anthracene 97%
9-(Naphthalen-1-yl)-10-(4-(naphthalen-2-yl)phenyl)anthracene

Precaución: for Research Use Only

Formula: C₄₀H₂₆
MW: 506.6300 g/mol
MDL Number: MFCD26388421
CAS Number: 1026768-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR