[2,2'-Bipyridine]-6,6'-dicarbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A1002573-250MG
A1002573-5G
A1002573-100MG
76645-954EA
14
USD
76645-954
76645-956
77619-486
[2,2'-Bipyridine]-6,6'-dicarbaldehyde 97%
[2,2'-Bipyridine]-6,6'-dicarbaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₈N₂O₂ MW: 212.2000 g/mol |
MDL Number:
MFCD06771787 CAS Number: 49669-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...