[2,2'-Bipyridine]-6,6'-diyldimethanol 97%

Supplier: Ambeed
Warning

A1002560-1G A1002560-250MG A1002560-5G A1002560-100MG
76645-942EA 109.28 USD
76645-942 76645-944 76645-946 77619-484
[2,2'-Bipyridine]-6,6'-diyldimethanol 97%
[2,2'-Bipyridine]-6,6'-diyldimethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.2359 g/mol
MDL Number: MFCD30738087
CAS Number: 74065-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR