(1R,2R)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride 97%

Supplier: Ambeed
Warning

A1323482-1G A1323482-250MG A1323482-100MG
76657-856EA 369 USD
76657-856 76657-858 77867-256
(1R,2R)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride 97%
(1R,2R)-N1,N1-Dimethyl-1,2-diphenylethane-1,2-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₂Cl₂N₂
CAS Number: 2635339-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR