Di-tert-butyl(methyl)phosphonium tetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: (t-Bu)2PMeHBF4, Ditert-butyl(methyl)phosphonium tetrafluoroborate, Bis(1,1-dimethylethyl)methylphosphine tetrafluoroborate

A1002190-1G A1002190-25G A1002190-5G
76645-924EA 30 USD
76645-924 76645-926 76645-928
Di-tert-butyl(methyl)phosphonium tetrafluoroborate 98%
Di-tert-butyl(methyl)phosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₉H₂₂BF₄P
MW: 248.05 g/mol
MDL Number: MFCD03840579
CAS Number: 479094-62-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR