(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride 98%

Supplier: Ambeed
Danger

A1002111-1G A1002111-25G A1002111-5G A1002111-250MG
76645-918EA 159.14 USD
76645-918 76645-920 76645-922 77570-008
(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride 98%
(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride

Precaución: for Research Use Only

Formula: C₈H₈F₂N₂O₅S₂
MW: 314.2863 g/mol
MDL Number: MFCD31625434
CAS Number: 2172794-56-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR