2-(Phenyliodonio)benzoate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Water 4-7%

A1002043-5G A1002043-25G
76645-916EA 52 USD
76645-916 77782-693
2-(Phenyliodonio)benzoate 98% stabilized
2-(Phenyliodonio)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₉IO₂
MDL Number: MFCD00032129
CAS Number: 1488-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR