RMC-4550 98+%

Supplier: Ambeed
Warning

Synonyms: (3-((3S,4S)-4-Amino-3-methyl-2-oxa-8-azaspiro[4.5]decan-8-yl)-6-(2,3-dichlorophenyl)-5-methylpyrazin-2-yl)methanol, RMC4550

A1001786-10MG A1001786-1MG A1001786-5MG
76645-910EA 157.02 USD
76645-910 76645-912 76645-914
RMC-4550 98+%
RMC-4550

Precaución: for Research Use Only

Formula: C₂₁H₂₆Cl₂N₄O₂
MW: 437.36 g/mol
CAS Number: 2172651-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR