(2-(4-((2'-Chloro-4',4'-difluoro-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-1-yl)methyl)-3-methyl-1H-pyrazol-1-yl)pyridin-3-yl)methanol (2R,3R)-2,3-dihydroxysuccinate 98+%

Supplier: Ambeed
Warning

Synonyms: LY2940094 tartrate

A1001766-1MG A1001766-25MG
76645-902EA 72.15 USD
76645-902 76645-904
(2-(4-((2'-Chloro-4',4'-difluoro-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-1-yl)methyl)-3-methyl-1H-pyrazol-1-yl)pyridin-3-yl)methanol (2R,3R)-2,3-dihydroxysuccinate 98+%
(2-(4-((2'-Chloro-4',4'-difluoro-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-1-yl)methyl)-3-methyl-1H-pyrazol-1-yl)pyridin-3-yl)methanol (2R,3R)-2,3-dihydroxysuccinate

Precaución: for Research Use Only

Formula: C₂₆H₂₉ClF₂N₄O₈S
MW: 631.05 g/mol
CAS Number: 1307245-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR