4,7-Diazaspiro[2.5]octan-8-one 97%

Supplier: Ambeed
Warning

A1001289-100MG A1001289-250MG
76645-880EA 118.83 USD
76645-880 77619-474
4,7-Diazaspiro[2.5]octan-8-one 97%
4,7-Diazaspiro[2.5]octan-8-one

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.1564 g/mol
MDL Number: MFCD07373478
CAS Number: 907973-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR