IR 783 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 4-(2-(2-(2-chloro-3-(2-(3,3-dimethyl-1-(4-sulphonatobutyl)-3H-indol-1-ium-2-yl)vinyl)cyclohex-2-en-1-ylidene)ethylidene)-3,3-dimethylindolin-1-yl)butane-1-sulphonate, Sodium 4-[2-[2-[2-Chloro-3-[2-[3,3-dimethyl-1-(4-sulphonatobutyl)indol-1-ium-2-yl]ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethylindol-1-yl]butane-1-sulphonate, 2-[2-[2-Chloro-3-[2-[1,3-dihydro-3,3-dimethyl-1-(4-sulphobutyl)-2H-indol-2-ylidene]ethylidene]-1-cyclohexen-1-yl]ethenyl]-3,3-dimethyl-1-(4-sulphobutyl)-3H-indolium Inner salt sodium salt

A1001251-100MG A1001251-1MG A1001251-1G A1001251-250MG
76645-876EA 26.53 USD
76645-876 76645-878 77569-994 77595-630
IR 783 98%
IR 783

Precaución: for Research Use Only

Formula: C₃₈H₄₆ClN₂NaO₆S₂
MW: 749.35 g/mol
MDL Number: MFCD03093208
CAS Number: 115970-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR