2-Isopropylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropyl-1H-benzimidazole, 2-Isopropyl-1H-benzo[d]imidazole

A118100-1G A118100-250MG A118100-25G A118100-5G
76657-832EA 11.67 USD
76657-832 76657-834 76657-836 76657-838
2-Isopropylbenzimidazole 98%
2-Isopropylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
MDL Number: MFCD00051759
CAS Number: 5851-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR