4-Pentylphenyl 4-(octyloxy)benzoate 97%

Supplier: Ambeed
Warning

A1001225-1G A1001225-25G A1001225-5G
76645-866EA 7 USD
76645-866 77782-681 77782-682
4-Pentylphenyl 4-(octyloxy)benzoate 97%
4-Pentylphenyl 4-(octyloxy)benzoate

Precaución: for Research Use Only

Formula: C₂₆H₃₆O₃
MDL Number: MFCD10565652
CAS Number: 50649-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR