tert-Butyl (4-bromo-2,6-difluorobenzyl)carbamate 97%

Supplier: Ambeed
Warning

A1001055-100MG A1001055-250MG A1001055-1G
76645-862EA 12 USD
76645-862 76645-864 77595-628
tert-Butyl (4-bromo-2,6-difluorobenzyl)carbamate 97%
tert-Butyl (4-bromo-2,6-difluorobenzyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrF₂NO₂
MDL Number: MFCD32011799
CAS Number: 1402673-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR