4-Propylbenzoic acid 98%

Supplier: Ambeed
Warning

A104217-100G A104217-10G A104217-25G A104217-500G
76657-824EA 41.38 USD
76657-824 76657-826 76657-828 76657-830
4-Propylbenzoic acid 98%
4-Propylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00013996
CAS Number: 2438-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR