Dinaphtho[2,1-b:1',2'-d]furan 97%

Supplier: Ambeed
Warning

A1000911-1G A1000911-5G
76645-856EA 47 USD
76645-856 77782-678
Dinaphtho[2,1-b:1',2'-d]furan 97%
Dinaphtho[2,1-b:1',2'-d]furan

Precaución: for Research Use Only

Formula: C₂₀H₁₂O
MDL Number: MFCD30833908
CAS Number: 194-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR