N,9,9-Triphenyl-9H-fluoren-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylamino-9,9-diphenyl-9H-fluorene

A1000905-100MG A1000905-1G A1000905-250MG
76645-850EA 14.86 USD
76645-850 76645-852 76645-854
N,9,9-Triphenyl-9H-fluoren-2-amine 97%
N,9,9-Triphenyl-9H-fluoren-2-amine

Precaución: for Research Use Only

Formula: C₃₁H₂₃N
MW: 409.53 g/mol
MDL Number: MFCD30470208
CAS Number: 860465-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR