Abt-724 97%

Supplier: Ambeed
Warning

Synonyms: 2-((4-(Pyridin-2-yl)piperazin-1-yl)methyl)-1H-benzo[d]imidazole

A1000642-10MG A1000642-1MG A1000642-25MG A1000642-50MG A1000642-5MG
76645-836EA 29 USD
76645-836 76645-838 76645-840 76645-842 76645-844
Abt-724 97%
Abt-724

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₅
MW: 293.37 g/mol
MDL Number: MFCD05262347
CAS Number: 70006-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR