Chlorobenzene 99.9% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
44565-0010
445651000
TS44565-0010EA
106
USD
TS44565-0010
TS445651000
Chlorobenzene 99.9% for HPLC
Chlorobenzene
Formula:
C₆H₅Cl Boiling Pt: 131 °C (760 mmHg) Melting Pt: –45 °C Density: 1.1075 g/cm³ Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 UN: 1134 ADR: 3,III |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.1 % |
GC | ≥99.85 % |
UV | at 360 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...