Chlorobenzene, extra dry over Molecular Sieve 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

44300-1000 443000010
TS44300-1000EA 29.83 USD
TS44300-1000 TS443000010
Chlorobenzene, extra dry over Molecular Sieve 99.5%, AcroSeal™
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 131 °C (760 mmHg)
Melting Pt: –45 °C
Density: 1.1075 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
UN: 1134
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR