Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
Supplier: Thermo Scientific Chemicals
|
Danger
|
42929-1000
429295000
429290025
TS42929-1000EA
56
USD
TS42929-1000
TS429295000
TS429290025
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
Amberlite® IRN-78, Ion exchange resin cationic type (OH-)
Storage Temperature:
Ambient |
MDL Number:
MFCD00145822 CAS Number: 11128-95-3 |
Specification Test Results
Appearance | Yellow to amber Spherical beads |
Harmonic mean size | 0.580 to 0.680 mm |
Uniformity coefficient | ≤1.20 |
Moisture holding capacity | 54.0 to 60.0 % (OH) |
Total volume capacity | ≥1.10 eq/l (OH) as is |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...