Lithium tetrahydroborate 10% (w/w) 4 M in THF, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithium tetrahydridoborate, Lithium borohydride
42888-8000
428881000
TS42888-8000EA
633
USD
TS42888-8000
TS428881000
Lithium tetrahydroborate 10% (w/w) 4 M in THF, AcroSeal™
Lithium tetrahydroborate
Formula:
H₄BLi Density: 0.89 g/cm³ Flash Pt: –21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011088 CAS Number: 16949-15-8 UN: 3399 ADR: 4.3,I |
Specification Test Results
Appearance | Hazy colorless to grey Solution |
Strength | 3.8 to 4.3 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...