Dibutyl ether, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-n-butyl ether

39691-1000 396910010 396910025
TS39691-1000EA 51 USD
TS39691-1000 TS396910010 TS396910025
Dibutyl ether, extra dry 99+%, AcroSeal™
Dibutyl ether
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 141
Melting Pt: -95
Density: 0.768 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009461
CAS Number: 142-96-1
UN: 1149
ADR: 3 III
Merck Index: 13,01568

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR