Trimethylamine 1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

38837-1000 388378000
TS38837-1000EA 171 USD
TS38837-1000 TS388378000
Trimethylamine 1 M in THF, AcroSeal™
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Density: 0.85 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 1 M Exact strength on the CoA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR