Cellulose, microcrystalline ∼ Ø 50 µm

Supplier: Thermo Scientific Chemicals

Synonyms: Microcrystallinecellulose

38724-0050 387240010 387242500
TS38724-0050EA 339 USD
TS38724-0050 TS387240010 TS387242500
Cellulose, microcrystalline ∼ Ø 50 µm
Cellulose
Formula: (C₁₂H₂₂O₁₁)n
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.5 %
Particle size ≥50 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR