Trimethylamine 50% (w/w) 7.3 M in aqueous solution, pure

Supplier: Thermo Scientific Chemicals
Danger

37078-5000 370780100 370780010 370780025
TS37078-5000EA 46 USD
TS37078-5000 TS370780100 TS370780010 TS370780025
Trimethylamine 50% (w/w) 7.3 M in aqueous solution, pure
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 30 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.86 g/cm³
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 48 to 52 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR