Hydrochloric acid 4 N in 1,4-dioxane/water

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

36652-0025 366521000
TS36652-0025EA 750 USD
TS36652-0025 TS366521000
Hydrochloric acid 4 N in 1,4-dioxane/water
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.1 g/cm³
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 3.95 to 4.40 N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR