Dichloromethane stabilised for residue analysis, for pesticide residue analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methylene chloride

Stabilization: Stabilised with amylene

32660-0040 326600010 326600025
TS32660-0040EA 239 USD
TS32660-0040 TS326600010 TS326600025
Dichloromethane stabilised for residue analysis, for pesticide residue analysis
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.325 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (K.F.)
GC (ECD) Passes test, (FID) Passes test
Stabilizer Approx. 50 ppm amylene
Residue after evaporation ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR