Ammonia solution 25% in water, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

25521-0010 255210025
TS25521-0010EA 61 USD
TS25521-0010 TS255210025
Ammonia solution 25% in water, Extra Pure
Ammonia solution
Formula: NH₃
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.91 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl ≥25 % (As NH3)
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR