L(+)-Sodium lactate 60% (w/w) in water

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt

25030-0010 250300050
TS25030-0010EA 163 USD
TS25030-0010 TS250300050
L(+)-Sodium lactate 60% (w/w) in water
L(+)-Sodium lactate
Formula: C₃H₅NaO₃
MW: 112.06 g/mol
Density: 1.33 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066576
CAS Number: 867-56-1
EINECS: 212-762-3

Order Now

Specification Test Results

Appearance Clear viscous liquid
Refractive index 1.4220 to 1.4250 (20 °C, 589 nm)
Density (20 °C) 1.32 to 1.34 g/ml
Color scale ≤50 APHA
pH 6.5 to 8.5
Titration with HCl 59 to 61 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) 12.1 to 12.6 %
Heavy metals (as Pb) ≤10 ppm
Isomer ≥97 % L(+) (Stereochemical purity)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR