2-Pentanone 99+%, AcroSeal™, purified by redistillation

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl Propyl Ketone

22413-1000 224130010
TS22413-1000EA 44 USD
TS22413-1000 TS224130010
2-Pentanone 99+%, AcroSeal™, purified by redistillation
2-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 100…103 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.809 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
UN: 1249
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20 °C, 589 nm)
GC ≥99.0 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR