Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder

Supplier: Thermo Scientific Chemicals
Danger

20623-1000 206230025 206235000
TS20623-1000EA 80 USD
TS20623-1000 TS206230025 TS206235000
Nickel aluminium alloy according to Raney (Al-Ni) 50/50, powder
Nickel aluminium alloy according to Raney
Formula: Ni:Al;₅₀:₅₀%
Density: 3.46
Storage Temperature: Ambient
MDL Number: MFCD00167392
CAS Number: 12635-27-7
UN: 3208
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Grey to grey-black Powder
Aluminium (Al) 47.0 to 52.0 %
Nickel (Ni) 48.0 to 53.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR