Molecular sieve A5 (0.5 nm, 5 Å) 8 --> 12 mesh

Supplier: Thermo Scientific Chemicals

Synonyms: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets

19729-5000 197290050
TS19729-5000EA 85 USD
TS19729-5000 TS197290050
Molecular sieve A5 (0.5 nm, 5 Å) 8 --> 12 mesh
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now

Specification Test Results

Appearance Beige to light brown Beads
Particle size 8 to 12 mesh
Bulk density 690 to 790 g/l
Residual water ≤1.5 % (550 °C, 2h)
Water adsorption ≥21 % (50%rH, 20 °C, 24h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR