Borane dimethyl sulfide complex 2 mol/l in THF, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
18507-1000
185078000
TS18507-1000EA
83
USD
TS18507-1000
TS185078000
Borane dimethyl sulfide complex 2 mol/l in THF, AcroSeal™
Borane dimethyl sulphide complex
Formula:
C₂H₉BS Flash Pt: –17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013189 CAS Number: 13292-87-0 EINECS: 236-313-6 UN: 3399 ADR: 4.3,I |
Specification Test Results
Appearance | Clear colorless to light yellow Solution |
Strength | 1.8 to 2.2 M |
Loss on drying | 4 % to 9 % (1 g, 105 °C) (vacuum) |
HPLC | ≥97.5 % |
Specific optical rotation | (20 °C, 589 nm) (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...