Polyvinyl alcohol, M.W. ∼ 50,000-85,000 98.0-98.8 %, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

18338-1000 183380010 183380025
TS18338-1000EA 27 USD
TS18338-1000 TS183380010 TS183380025
Polyvinyl alcohol, M.W. ∼ 50,000-85,000 98.0-98.8 %, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to cream Granular powder
Infrared spectrum Conforms
Viscosity 9 to 11 cP (4 % in water)
Degree of hydrolysis 98.0 to 98.8 mol %
Ash ≤1.2 % (as Na2O)
pH 5.0 to 7.0 (4 % in water)
Volatile matter ≤5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR